C12H316CF190 Chemcia Scientific, LLC

(4-Fluoro-pyridin-2-yl)-methyl-amine

  • MDL number MFCD11848520
  • Cas Number : 1260832-12-9
  • Molecular Weight : 126.13449

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7047

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7047
Availability details for
Quantity
  • Availability
  • Add to Cart