C12H316CF193 Chemcia Scientific, LLC

2-Amino-5-bromo-4-chloro-benzonitrile

  • MDL number MFCD08752639
  • Cas Number : 671795-60-1
  • Molecular Weight : 231.48033

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7052

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7052
Availability details for
Quantity
  • Availability
  • Add to Cart