C12H316CF1AF Chemcia Scientific, LLC

4-Fluoro-5-methoxy-2-nitro-benzoic acid methyl ester

  • MDL number MFCD20921594
  • Cas Number : 159768-50-0
  • Molecular Weight : 229.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4-fluoro-5-methoxy-2-nitrobenzoate
Inchi Code 1S/C9H8FNO5/c1-15-8-3-5(9(12)16-2)7(11(13)14)4-6(8)10/h3-4H,1-2H3
Inchi Key QLBGQKDRKJSSFI-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=7226

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7226
Availability details for
Quantity
  • Availability
  • Add to Cart