C12H316CF1B3 Chemcia Scientific, LLC

2-Bromomethyl-4-chloro-benzonitrile

  • MDL number MFCD11847520
  • Cas Number : 50712-67-9
  • Molecular Weight : 230.49275

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7115

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7115
Availability details for
Quantity
  • Availability
  • Add to Cart