C12H316CF1B4 Chemcia Scientific, LLC

4-Chloro-2-methyl-5-nitro-benzonitrile

  • Cas Number : 178448-14-1
  • Molecular Weight : 196.59425

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7117

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7117
Availability details for
Quantity
  • Availability
  • Add to Cart