C12H316CF1B6 Chemcia Scientific, LLC

4-Chloro-5-iodo-2-methyl-benzoic acid methyl ester

  • MDL number MFCD30180148
  • Cas Number : 1399182-04-7
  • Molecular Weight : 310.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4-chloro-5-iodo-2-methylbenzoate
Inchi Code 1S/C9H8ClIO2/c1-5-3-7(10)8(11)4-6(5)9(12)13-2/h3-4H,1-2H3
Inchi Key ACDLTDLWSKUIJT-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=7128

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7128
Availability details for
Quantity
  • Availability
  • Add to Cart