C12H316CF1B8 Chemcia Scientific, LLC

5-Chloro-4-fluoro-2-methoxy-benzoic acid

  • MDL number MFCD22573011
  • Cas Number : 1555667-91-8
  • Molecular Weight : 204.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-chloro-4-fluoro-2-methoxybenzoic acid
Inchi Code 1S/C8H6ClFO3/c1-13-7-3-6(10)5(9)2-4(7)8(11)12/h2-3H,1H3,(H,11,12)
Inchi Key BSYFUUVHTCUXKE-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=7144

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7144
Availability details for
Quantity
  • Availability
  • Add to Cart