C12H316CF1D2 Chemcia Scientific, LLC

2-Methyl-6-nitro-2,3-dihydro-isoindol-1-one

  • MDL number MFCD20226157
  • Cas Number : 564467-91-0
  • Molecular Weight : 192.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-methyl-6-nitro-1-isoindolinone
Inchi Code 1S/C9H8N2O3/c1-10-5-6-2-3-7(11(13)14)4-8(6)9(10)12/h2-4H,5H2,1H3
Inchi Key ZAEFFIFTXQYMPA-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=7198

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7198
Availability details for
Quantity
  • Availability
  • Add to Cart