C12H316CF1F9 Chemcia Scientific, LLC

1-(3-Methyl-4-nitro-phenyl)-ethanone

  • MDL number MFCD01823165
  • Cas Number : 96784-58-6
  • Molecular Weight : 179.17698

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7326

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7326
Availability details for
Quantity
  • Availability
  • Add to Cart