C12H316CF1FF Chemcia Scientific, LLC

1-(5-Amino-2-fluoro-4-hydroxy-phenyl)-ethanone

  • MDL number MFCD30180181
  • Cas Number : 1613720-25-4
  • Molecular Weight : 169.15686

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7972

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7972
Availability details for
Quantity
  • Availability
  • Add to Cart