C12H316CF202 Chemcia Scientific, LLC

6-Fluoro-5-methoxy-benzothiazol-2-ylamine

  • MDL number MFCD20714054
  • Cas Number : 1344684-78-1
  • Molecular Weight : 198.22019

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7661

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7661
Availability details for
Quantity
  • Availability
  • Add to Cart