C12H316CF203 Chemcia Scientific, LLC

2-Chloro-6-fluoro-5-methoxy-benzothiazole

  • MDL number MFCD30180184
  • Cas Number : 1344684-79-2
  • Molecular Weight : 217.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-chloro-6-fluoro-5-methoxybenzo[d]thiazole
Inchi Code 1S/C8H5ClFNOS/c1-12-6-3-5-7(2-4(6)10)13-8(9)11-5/h2-3H,1H3
Inchi Key YFOKOXIRXBOQGF-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=7660

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7660
Availability details for
Quantity
  • Availability
  • Add to Cart