C12H316CF204 Chemcia Scientific, LLC

1-Bromo-5-chloro-4-fluoro-2-nitro-benzene

  • MDL number MFCD12026230
  • Cas Number : 960000-99-1
  • Molecular Weight : 254.44374

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7321

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7321
Availability details for
Quantity
  • Availability
  • Add to Cart