C12H316CF206 Chemcia Scientific, LLC

3-Amino-4-methyl-2-nitro-benzoic acid

  • MDL number MFCD01098202
  • Cas Number : 37901-90-9
  • Molecular Weight : 196.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-amino-4-methyl-2-nitrobenzoic acid
Inchi Code 1S/C8H8N2O4/c1-4-2-3-5(8(11)12)7(6(4)9)10(13)14/h2-3H,9H2,1H3,(H,11,12)
Inchi Key JLWAVVAJFWDXEF-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=7309

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7309
Availability details for
Quantity
  • Availability
  • Add to Cart