C12H316CF207 Chemcia Scientific, LLC

3-Bromo-4-methyl-2-nitro-benzoic acid

  • MDL number MFCD28745206
  • Cas Number : 320740-32-7
  • Molecular Weight : 260.04532

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7308

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7308
Availability details for
Quantity
  • Availability
  • Add to Cart