C12H316CF208 Chemcia Scientific, LLC

2-Amino-3-bromo-4-methyl-benzoic acid

  • MDL number MFCD12755819
  • Cas Number : 320740-35-0
  • Molecular Weight : 230.06246

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0-5*C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7307

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7307
Availability details for
Quantity
  • Availability
  • Add to Cart