C12H316CF20B Chemcia Scientific, LLC

3-Methoxy-4-methyl-2-nitro-benzoic acid methyl ester

  • MDL number MFCD12407814
  • Cas Number : 128450-32-8
  • Molecular Weight : 225.20287

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7300

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7300
Availability details for
Quantity
  • Availability
  • Add to Cart