C12H316CF20F Chemcia Scientific, LLC

7-Bromo-1H-cinnolin-4-one

  • MDL number MFCD28040482
  • Cas Number : 1414956-67-4
  • Molecular Weight : 225.04585

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7350

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7350
Availability details for
Quantity
  • Availability
  • Add to Cart