C12H316CF216 Chemcia Scientific, LLC

7-Bromo-1H-benzoimidazole

  • MDL number MFCD11973696
  • Cas Number : 83741-35-9
  • Molecular Weight : 197.0353

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Grey Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7378

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7378
Availability details for
Quantity
  • Availability
  • Add to Cart