C12H316CF21F Chemcia Scientific, LLC

5-Chloro-3-iodo-2H-pyrazolo[3,4-c]pyridine

  • MDL number MFCD30180209
  • Molecular Weight : 279.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-chloro-3-iodo-2H-pyrazolo[3,4-c]pyridine
Inchi Code 1S/C6H3ClIN3/c7-5-1-3-4(2-9-5)10-11-6(3)8/h1-2H,(H,10,11)
Inchi Key KAGKORFWYBVTEW-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=7400

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7400
Availability details for
Quantity
  • Availability
  • Add to Cart