C12H316CF225 Chemcia Scientific, LLC

5-Fluoro-1H-pyrrolo[2,3-c]pyridine-2-carboxylic acid ethyl ester

  • MDL number MFCD13196661
  • Cas Number : 920978-84-3
  • Molecular Weight : 208.19383

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7415

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7415
Availability details for
Quantity
  • Availability
  • Add to Cart