C12H316CF227 Chemcia Scientific, LLC

3-Bromo-6-methoxy-2-methyl-pyridine 1-oxide

  • Cas Number : 269058-50-6
  • Molecular Weight : 218.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-bromo-6-methoxy-2-methylpyridine 1-oxide
Inchi Code 1S/C7H8BrNO2/c1-5-6(8)3-4-7(11-2)9(5)10/h3-4H,1-2H3
Inchi Key GKHNPEMIHCUEHI-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=7431

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7431
Availability details for
Quantity
  • Availability
  • Add to Cart