C12H316CF22F Chemcia Scientific, LLC

3-Bromo-2-methoxymethoxy-benzonitrile

  • Molecular Weight : 242.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-bromo-2-(methoxymethoxy)benzonitrile
Inchi Code 1S/C9H8BrNO2/c1-12-6-13-9-7(5-11)3-2-4-8(9)10/h2-4H,6H2,1H3
Inchi Key LTYYNDPBWWHMNN-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Light Yellow Liquid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=7469

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7469
Availability details for
Quantity
  • Availability
  • Add to Cart