C12H316CF23C Chemcia Scientific, LLC

1-(4-Bromo-2-chloro-5-fluoro-phenyl)-ethanone

  • MDL number MFCD28009840
  • Cas Number : 1435805-96-1
  • Molecular Weight : 251.48385

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7506

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7506
Availability details for
Quantity
  • Availability
  • Add to Cart