C12H316CF240 Chemcia Scientific, LLC

2-Iodo-4-nitro-benzamide

  • MDL number MFCD13192770
  • Cas Number : 89677-75-8
  • Molecular Weight : 292.0339

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7515

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7515
Availability details for
Quantity
  • Availability
  • Add to Cart