C12H316CF241 Chemcia Scientific, LLC

2-Iodo-4-nitro-benzonitrile

  • MDL number MFCD18397969
  • Cas Number : 300627-43-4
  • Molecular Weight : 274.01856

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7516

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7516
Availability details for
Quantity
  • Availability
  • Add to Cart