C12H316CF243 Chemcia Scientific, LLC

4-Iodo-2-trifluoromethyl-benzoic acid methyl ester

  • MDL number MFCD09881758
  • Cas Number : 954815-14-6
  • Molecular Weight : 330.04657

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Yellow Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7519

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7519
Availability details for
Quantity
  • Availability
  • Add to Cart