C12H316CF245 Chemcia Scientific, LLC

4-Iodo-2-trifluoromethyl-benzaldehyde

  • MDL number MFCD15528741
  • Cas Number : 1007579-67-0
  • Molecular Weight : 300.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-iodo-2-(trifluoromethyl)benzaldehyde
Inchi Code 1S/C8H4F3IO/c9-8(10,11)7-3-6(12)2-1-5(7)4-13/h1-4H
Inchi Key KKFCXRKONPDJPV-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=7521

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7521
Availability details for
Quantity
  • Availability
  • Add to Cart