C12H316CF247 Chemcia Scientific, LLC

3-Amino-6-chloro-thieno[3,2-b]pyridine-2-carboxylic acid methyl ester

  • Cas Number : 208994-12-1
  • Molecular Weight : 242.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 3-amino-6-chlorothieno[3,2-b]pyridine-2-carboxylate
Inchi Code 1S/C9H7ClN2O2S/c1-14-9(13)8-6(11)7-5(15-8)2-4(10)3-12-7/h2-3H,11H2,1H3
Inchi Key CJTHRTORVFJNHH-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=7550

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7550
Availability details for
Quantity
  • Availability
  • Add to Cart