C12H316CF248 Chemcia Scientific, LLC

4-Amino-5-iodo-2-methyl-benzoic acid

  • MDL number MFCD30180226
  • Cas Number : 1934511-55-3
  • Molecular Weight : 277.06286

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7552

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7552
Availability details for
Quantity
  • Availability
  • Add to Cart