C12H316CF24B Chemcia Scientific, LLC

(4-Iodo-2-nitro-phenyl)-methyl-amine

  • Cas Number : 1260791-09-0
  • Molecular Weight : 278.05044

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Orange Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7573

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7573
Availability details for
Quantity
  • Availability
  • Add to Cart