C12H316CF24C Chemcia Scientific, LLC

5-Iodo-1H-benzoimidazole

  • MDL number MFCD06659631
  • Cas Number : 78597-27-0
  • Molecular Weight : 244.0357

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7574

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7574
Availability details for
Quantity
  • Availability
  • Add to Cart