C12H316CF250 Chemcia Scientific, LLC

3-Fluoro-4-iodo-2-methyl-phenylamine

  • Cas Number : 329927-16-4
  • Molecular Weight : 251.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-fluoro-4-iodo-2-methylaniline
Inchi Code 1S/C7H7FIN/c1-4-6(10)3-2-5(9)7(4)8/h2-3H,10H2,1H3
Inchi Key NFJMWGRBWAJRLD-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=7581

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7581
Availability details for
Quantity
  • Availability
  • Add to Cart