C12H316CF252 Chemcia Scientific, LLC

5-Bromo-4-fluoro-2H-indazole

  • MDL number MFCD28392873
  • Cas Number : 1082041-85-7
  • Molecular Weight : 215.02573

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7583

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7583
Availability details for
Quantity
  • Availability
  • Add to Cart