C12H316CF253 Chemcia Scientific, LLC

4-Bromo-2-tert-butyl-phenylamine

  • MDL number MFCD09965991
  • Cas Number : 850012-44-1
  • Molecular Weight : 228.13378

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Brown Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7594

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7594
Availability details for
Quantity
  • Availability
  • Add to Cart