C12H316CF254 Chemcia Scientific, LLC

2-tert-Butyl-5-nitro-phenylamine

  • Cas Number : 103392-84-3
  • Molecular Weight : 194.23528

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7595

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7595
Availability details for
Quantity
  • Availability
  • Add to Cart