C12H316CF257 Chemcia Scientific, LLC

(2-tert-Butyl-phenyl)-methyl-amine

  • MDL number MFCD11934511
  • Cas Number : 109932-97-0
  • Molecular Weight : 163.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-tert-butyl-N-methylaniline
Inchi Code 1S/C11H17N/c1-11(2,3)9-7-5-6-8-10(9)12-4/h5-8,12H,1-4H3
Inchi Key DHQJBOWJBHNPIF-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Brown Liquid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=7598

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7598
Availability details for
Quantity
  • Availability
  • Add to Cart