C12H316CF258 Chemcia Scientific, LLC

5-Amino-2-tert-butyl-phenol

  • MDL number MFCD16996855
  • Cas Number : 873055-35-7
  • Molecular Weight : 165.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-amino-2-tert-butylphenol
Inchi Code 1S/C10H15NO/c1-10(2,3)8-5-4-7(11)6-9(8)12/h4-6,12H,11H2,1-3H3
Inchi Key WRTDVWHCSGVIOT-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=7599

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7599
Availability details for
Quantity
  • Availability
  • Add to Cart