C12H316CF262 Chemcia Scientific, LLC

2-Bromo-4-chloro-5-nitro-benzoic acid

  • MDL number MFCD30180236
  • Molecular Weight : 280.46326

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7621

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7621
Availability details for
Quantity
  • Availability
  • Add to Cart