C12H316CF263 Chemcia Scientific, LLC

5-Fluoro-4-iodo-2-nitro-phenylamine

  • MDL number MFCD21686787
  • Cas Number : 1219741-86-2
  • Molecular Weight : 282.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-fluoro-4-iodo-2-nitroaniline
Inchi Code 1S/C6H4FIN2O2/c7-3-1-5(9)6(10(11)12)2-4(3)8/h1-2H,9H2
Inchi Key VYUKTTWGCYDBIV-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=7623

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7623
Availability details for
Quantity
  • Availability
  • Add to Cart