C12H316CF26A Chemcia Scientific, LLC

3-Chloro-6-hydroxy-[1,5]naphthyridine-4-carboxylic acid

  • MDL number MFCD30180237
  • Molecular Weight : 224.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Inchi Code 1S/C9H5ClN2O3/c10-4-3-11-5-1-2-6(13)12-8(5)7(4)9(14)15/h1-3H,(H,12,13)(H,14,15)
Inchi Key IGRGCHAVRREDRX-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Yellow Solid
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=7637
Availability details for
Quantity
  • Availability
  • Add to Cart