C12H316CF26B Chemcia Scientific, LLC

3-Chloro-6-hydroxy-7-methyl-[1,5]naphthyridine-4-carboxylic acid

  • MDL number MFCD30180238
  • Molecular Weight : 238.63189

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7638

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7638
Availability details for
Quantity
  • Availability
  • Add to Cart