C12H316CF28A Chemcia Scientific, LLC

(5-Chloro-1H-pyrrolo[2,3-c]pyridin-2-yl)-methanol

  • MDL number MFCD22045074
  • Cas Number : 867036-42-8
  • Molecular Weight : 182.61079

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7703

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7703
Availability details for
Quantity
  • Availability
  • Add to Cart