C12H316CF292 Chemcia Scientific, LLC

2-Amino-5-iodo-3-methoxy-benzoic acid

  • MDL number MFCD28658370
  • Cas Number : 1616831-34-5
  • Molecular Weight : 293.06226

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7722

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7722
Availability details for
Quantity
  • Availability
  • Add to Cart