C12H316CF295 Chemcia Scientific, LLC

1H-Indazole-3,5-diamine hydrochloride

  • MDL number MFCD30478184
  • Cas Number : 19335-15-0
  • Molecular Weight : 184.62958

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7665

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7665
Availability details for
Quantity
  • Availability
  • Add to Cart