C12H316CF2B1 Chemcia Scientific, LLC

5-Amino-2-fluoro-phenol

  • MDL number MFCD13185335
  • Cas Number : 100367-48-4
  • Molecular Weight : 127.11922

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7773

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7773
Availability details for
Quantity
  • Availability
  • Add to Cart