C12H316CF2B7 Chemcia Scientific, LLC

2-Amino-5-chloro-3-fluoro-benzoic acid

  • MDL number MFCD11193641
  • Cas Number : 1028757-83-6
  • Molecular Weight : 189.5748

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7784

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7784
Availability details for
Quantity
  • Availability
  • Add to Cart