C12H316CF2B9 Chemcia Scientific, LLC

2-Amino-3-fluoro-benzaldehyde

  • MDL number MFCD12923269
  • Cas Number : 854538-94-6
  • Molecular Weight : 139.13037

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7786

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7786
Availability details for
Quantity
  • Availability
  • Add to Cart