C12H316CF2BA Chemcia Scientific, LLC

(5-Methoxy-imidazo[1,2-a]pyridin-2-yl)-methanol

  • MDL number MFCD30478199
  • Cas Number : 1783772-86-0
  • Molecular Weight : 178.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (5-methoxyimidazo[1,2-a]pyridin-2-yl)methanol
Inchi Code 1S/C9H10N2O2/c1-13-9-4-2-3-8-10-7(6-12)5-11(8)9/h2-5,12H,6H2,1H3
Inchi Key ODXKSCQYJLTXRX-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=7793

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7793
Availability details for
Quantity
  • Availability
  • Add to Cart