C12H316CF2BB Chemcia Scientific, LLC

5-Methoxy-imidazo[1,2-a]pyridine-2-carbaldehyde

  • MDL number MFCD22066376
  • Cas Number : 881841-58-3
  • Molecular Weight : 176.17631

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7794

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7794
Availability details for
Quantity
  • Availability
  • Add to Cart